Human Scrambled Jag1

Supplier: AnaSpec
AS-64239
103006-692EA 311.31 USD
103006-692
Human Scrambled Jag1
Proteins and Peptides
This peptide is scrambled sequence of JAG-1 (188-204), Jagged-1 (188-204), Notch Ligand, DSL Peptide, cat# 61298.Related Product: JAG-1 (188-204), Jagged-1 (188-204), Notch Ligand, DSL Peptide, cat# 61298.
Sequence: RCGPDCFDNYGRYKYCF
MW: 2107.5 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR