Human Protease Activated Receptor 1

Supplier: AnaSpec
AS-65479
103010-002EA 343.46 USD
103010-002
Human Protease Activated Receptor 1
Proteins and Peptides
This peptide is PAR-1 selective agonist displaying a high level of specificity to PAR-1 over PAR-2. The specificity of peptide was evaluated in cell-based calcium signaling assay using HEK293 cells. PAR-1 selective agonists can be used to study PAR-1 activation in vivo.
Sequence: AF(para - Fluoro)R - Cha - Cit - Y - NH2
MW: 883 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR