Human Beta-Amyloid (10-20)

Supplier: AnaSpec
AS-25365
102999-858EA 118.38 USD
102999-858
Human Beta-Amyloid (10-20)
Proteins and Peptides
A number of Aß fragments including Aß (10-20) enhances aggregation of Aß (1-40). All the Aß peptides that enhance aggregation contain either residues 17 to 20 or 30 to 35, indicating the importance of these regions for promoting aggregation of full-length Aß.
Sequence: YEVHHQKLVFF
Molecular Weight: 1446.7 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR