Human Beta-Amyloid (1-17)

Supplier: AnaSpec
AS-61955
103007-104EA 206.08 USD
103007-104
Human Beta-Amyloid (1-17)
Proteins and Peptides
This is beta-Amyloid peptide fragment derived from amino acids 1 to 17. This peptide was employed in the b-Amyloid solubility studies.
Sequence: DAEFRHDSGYEVHHQKL
Molecular Weight: 2068.2 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR