Testosterone ELISA Kit, Cayman Chemical

Supplier: Cayman Chemical Company

582701-480 582701-96
75816-940EA 1396.82 USD
75816-940 75816-942
Testosterone ELISA Kit, Cayman Chemical
Assays ELISAs

For the quantification of testosterone in serum, plasma, tissue culture supernatants and other sample matrices.


  • Measure testosterone in multiple sample types
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure testosterone levels down to 6 pg/mL
  • Incubation: 2 hours | Development: 60 – 90 minutes | Read: Colorimetric at 405 – 420 nm
  • Mid-point: 50% B/B0: 20 – 50 pg/mL


Testosterone is the prototypic and predominant circulating androgenic steroid. It plays a major role in the growth and function of many reproductive and non-reproductive tissues and organs including muscle, liver, and brain, directing the development of the male phenotype during embryogenesis and at puberty. Cayman's Testosterone ELISA Kit is a competitive assay that can be used for quantification of testosterone in serum, plasma, tissue.


Delivery information: Kit contains: Antiserum, Tracer, Standard, Buffers, Reagents, Plate, and Dye.


Packaging: Kit contains: Antiserum, Tracer, Standard, Buffers, Reagents, Plate, and Dye

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR