Anti-HA-tag Mouse Monoclonal Antibody [clone: 16B12]

Supplier: Enzo Life Sciences
ABS118-0500 ABS118-0200 ABS118-1000
76002-602EA 780.44 USD
76002-602 76002-604 76002-606
Anti-HA-tag Mouse Monoclonal Antibody [clone: 16B12]
Antibodies
Recommended Applications: IF, IHC, IP, WB

Species reactivity:

Type: Primary
Antigen: HA-tag
Clonality: Monoclonal
Clone: 16B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR