Human Recombinant Fc (human) IgG1 Control (from HEK293 cells)
Supplier: AdipoGen Life Sciences
CHI-HF-210IG1-C100
102981-690EA 456.92 USD
102981-690
Human Recombinant Fc (human) IgG1 Control (from HEK293 cells)
Proteins and Peptides
This product should be used as the control protein for activity tests of fusion proteins including human target proteins and human IgG-Fc domains.