Human CD54 (from CHO cells)
Supplier: AdipoGen Life Sciences
ANC-514-030
102513-286EA 750.46 USD
102513-286
Human CD54 (from CHO cells)
Proteins and Peptides
Human CD54 (ICAM-1) mediates cell adhesion by binding to the integrins CD11a/CD18 (LFA-1) and to CD11b/CD18 (Mac-1). CD54 expression on resting peripheral blood leukocytes is weak but is upregulated on activated T and B lymphocytes and on monocytes.