Human CD28 (from CHO cells)
Supplier: AdipoGen Life Sciences
ANC-508-020
102513-252EA 617.54 USD
102513-252
Human CD28 (from CHO cells)
Proteins and Peptides
Human CD28 is an important costimulatory molecule found on all CD4+ T cells and on about half of the CD8+T cells. T cell activities attributed to CD28 include prevention of anergy, induction of cytokine gene transcription, stabilization of cytokine mRNAs and activation of CD8+ cytotoxic T lymphocytes. Monovalency of the CD28 receptor for its natural ligands is essential to provide costimulation without inducing responses in the absense of TCR engagement.