Human CD137L (from CHO cells)
Supplier: AdipoGen Life Sciences
ANC-503-030
102513-230EA 750.46 USD
102513-230
Human CD137L (from CHO cells)
Proteins and Peptides
Human CD137L (4-1BB ligand) is a type II transmembrane protein constitutively expressed by monocytes, B cells and neuroblastoma cells. Binding of CD137 (4-1BB) to CD137L induces monocyte activation.