Human Recombinant VEGF 165 (from Insect Cells (Sf9))
Supplier: 0000027161
AG-40T-0045-C005 AG-40T-0045-C020
102980-402EA 332.31 USD
102980-402 102980-404
Human Recombinant VEGF 165 (from Insect Cells (Sf9))
Proteins and Peptides
Human vascular endothelial growth factor (VEGF 165) is produced as a homodimer. It binds to the FLT1/VEGFR1 and KDR/VEGFR2 receptors, heparan sulfate and heparin. VEGF 165 is a specific mitogen for vascular endothelial cells and a strong angiogenic factor in vivo. In addition to its action as a mitogen it is a potent vascular permeability factor (VPF) in vivo. It is also a chemoattractant molecule for monocytes and endothelial cells. It induces endothelial cell proliferation, promotes cell migration, inhibits apoptosis and induces permeabilization of blood vessels.