KU-0063794 ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: (5-(2-(cis-2,6-Dimethylmorpholino)-4-morpholinopyrido[2,3-d]pyrimidin-7-yl)-2-methoxyphenyl)methanol

CHM135-0005 CHM135-0010
76003-040EA 247.43 USD
76003-040 76003-042
KU-0063794 ≥99% (by HPLC)
KU-0063794

Highly selective and potent inhibitor against both mTORC1 and mTORC2.

Formula: C₂₅H₃₁N₅O₄
MDL Number: MFCD22666594
CAS Number: 938440-64-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbaab94ae033e136eadef91ba92e2ada5403fceb&vid=e597c3acaa10f607f47855133b6df09f72cd520a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR