27-Hydroxycholesterol ≥98% (by HPLC)

Supplier: Enzo Life Sciences

CHM102-0001 CHM102-0005
76002-956EA 73.03 USD
76002-956 76002-958
27-Hydroxycholesterol ≥98% (by HPLC)
27-Hydroxycholesterol

Cholesterol metabolite linked to breast cancer, neurodegenerative disease and metabolism.


27-Hydroxycholesterol (27-HC) is a metabolite resulting from the action of sterol 27-Hydroxylase on cholesterol. It is an endogenous selective estrogen receptor modulator and ligand for the liver X receptor (LXR). 27-Hydroxycholesterol plays a role in Alzheimer's disease, oxidative stress, metabolism, cell-signaling, and heart disease.


Soluble in DMSO (up to 10 mg/ml with warming) or in ethanol (up to 10 mg/ml). Insoluble in water.


Synonyms: 5,25R-​Cholest​en-​3β,​26-​diol, (25R)-26-Hydroxycholesterol

Formula: C₂₇H₄₆O₂
Storage Temperature: Freezer
CAS Number: 20380-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41246d5c0320d9bd08b6caa11cb7b09255159684&vid=bcab6e77cf6f634830fce6305eaad339d433c0de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR