Water, distilled
Supplier: ADIPOGEN CORP MS
Synonyms:
Dihydrogen oxide
IAX-900-002-LD15
IAX-900-002-L010
IAX-900-002-3010
102979-266EA
34.62
USD
102979-266
102979-268
102979-270
Water, distilled
Water
Formula:
H₂O MW: 18.02 g/mol Boiling Pt: 100 Melting Pt: 0 Storage Temperature: Refrigerator |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 Merck Index: 13,10098 |
Specification Test Results
Formulation | Liquid. 1.5ml (IAX-900-002-LD15) or 10ml (IAX-900-002-L010) sterile filtered double-distilled water (ddWater), without any additives. |
Endotoxin | <0.0002EU/µg, after repeated purification cycles over activated charcoal. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...