IPI-145 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
(S)-3-(1-((9H-Purin-6-yl)amino)ethyl)-8-chloro-2-phenylisoquinolin-1(2H)-one
SYN-1175-M001
SYN-1175-M005
SYN-1175-M010
SYN-1175-M050
102992-564EA
156.46
USD
102992-564
102992-566
102992-568
102992-570
IPI-145 ≥95%
IPI-145
IPI-145 is a potent, oral inhibitor of Class I phosphoinositide-3-kinase (PI3K)-delta and PI3K-gamma (IC(50) of 243 pM and 50nM), and is clinically active in patients with advanced hematological malignancies.
Formula:
C₂₂H₁₇ClN₆O MW: 416.9 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD15144635 CAS Number: 1201438-56-3 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Soluble in DMSO. Slightly soluble ( |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...