RG7422 95
Supplier: ADIPOGEN CORP MS
Synonyms:
(S)-1-(4-((2-(2-aminopyrimidin-5-yl)-7-methyl-4-morpholinothieno[3,2-d]pyrimidin-6-yl)methyl)piperazin-1-yl)-2-hydroxypropan-1-one, Apitolisib, GDC-0980
SYN-1150-M001
SYN-1150-M005
SYN-1150-M010
SYN-1150-M050
102992-384EA
149.54
USD
102992-384
102992-386
102992-388
102992-390
RG7422 95
RG7422
GDC-0980 (RG7422) is a selective, dual PI3 Kinase and mTOR Kinase inhibitor with IC(50) values of 5, 27, 7, and 14nM for PI3Kalpha-delta respectively. It inhibits mTOR with a Ki of 17nM.
Formula:
C₂₃H₃₀N₈O₃S MW: 498.6 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD20274520 CAS Number: 1032754-93-0 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Soluble in DMSO. Slightly soluble ( |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...