Ruxolitinib ≥95%

Supplier: ADIPOGEN CORP MS
Warning

SYN-1120-M001 SYN-1120-M005 SYN-1120-M010 SYN-1120-M050
102992-174EA 119.08 USD
102992-174 102992-176 102992-178 102992-180
Ruxolitinib ≥95%
Ruxolitinib

INCB018424 is a JAK inhibitor with an IC(50) values of 2.7nM and 4.5nM, respectively for JAK1 and JAK2.

Formula: C₁₇H₁₈N₆
Storage Temperature: Freezer
MDL Number: MFCD12031592
CAS Number: 941678-49-5

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR