Lenvatinib (E7080) ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
4-[3-chloro-4-(cyclopropylcarbamoylamino)phenoxy]-7-methoxyquinoline-6-carboxamide, Lenvatinib, 4-(3-Chloro-4-(3-cyclopropylureido)phenoxy)-7-methoxyquinoline-6-carboxamide
SYN-1038-M001
SYN-1038-M005
SYN-1038-M010
SYN-1038-M050
102991-542EA
134.31
USD
102991-542
102991-544
102991-546
102991-548
Lenvatinib (E7080) ≥95%
Lenvatinib (E7080)
E7080 is an orally active inhibitor of multiple receptor tyrosine kinases including VEGF, FGF and SCF receptors. It is most potent against VEGFR2(KDR)/VEGFR3(Flt-4), with IC(50) values of 4nM and 5.2nM respectively.
Formula:
C₂₁H₁₉ClN₄O₄ MW: 426.85 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD16038644 CAS Number: 417716-92-8 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...