n-Tetradecanoyl-L-homoserine lactone ≥97%

Supplier: ADIPOGEN CORP MS

Synonyms: C14-HSL, (S)-N-(2-Oxotetrahydrofuran-3-yl)tetradecanamide

CDX-T0144-M010 CDX-T0144-M050
102991-138EA 123.23 USD
102991-138 102991-140
n-Tetradecanoyl-L-homoserine lactone ≥97%
n-Tetradecanoyl-L-homoserine lactone

N-Tetradecanoyl-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. It appears later than shorter acyl chain AHLs in developing biofilms and, like other long chain AHLs, stimulates bacterial growth. C14-HSL also alters the proteolytic activity and enhances the migration of some strains of Proteus mirabilis. Induces violacein expression in a Chromobacterium violaceum mutant usually not able to produce homoserine lactones.

Formula: C₁₈H₃₃NO₃
MW: 311.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28899662
CAS Number: 202284-87-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (GC)
Appearance/Color White solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR