N-Boc-N′-succinyl-4,7,10-trioxa-1,13-tridecanediamine 95

Supplier: ADIPOGEN CORP MS
Warning

CDX-T0100-GG25 CDX-T0100-G025
102991-120EA 315.69 USD
102991-120 102991-122
N-Boc-N′-succinyl-4,7,10-trioxa-1,13-tridecanediamine 95
N-Boc-N′-succinyl-4,7,10-trioxa-1,13-tridecanediamine

Bifunctional linker for chemical synthesis.

Formula: C₁₉H₃₆N₂O₈
MW: 420.5 g/mol
Storage Temperature: Freezer
CAS Number: 250612-31-8

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Colourless viscous liquid.
Solubility Soluble in DMF.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR