Methoxyfenozide ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
Methoxyphenozide
CDX-M0133-M100
CDX-M0133-G001
102990-532EA
438.92
USD
102990-532
102990-530
Methoxyfenozide ≥98% (by HPLC)
Methoxyfenozide
Diacylhydrazine insecticide. Ecdysone agonist that mimics the biological function of the natural insect molting hormone 20-hydroxyecdysone, causing premature molting and interfering with the normal growth and development in insects. Ecdysteroid agonist are also used as ligands for gene expression and gene therapy research. Compound can be used as analytical reference material.
Formula:
C₂₂H₂₈N₂O₃ MW: 368.5 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03792750 CAS Number: 161050-58-4 UN: 3077 ADR: 9,III |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White powder. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...