1-(3-Isothiocyanatobenzyl)-4-[2-(3,4-dihydro-2H-1-benzopyran-6-yl)-5-oxazolyl] pyridinium bromide ≥90% (by HPCE)
Supplier: ADIPOGEN CORP MS
|
Warning
|
CDX-I0034-M025
CDX-I0034-M050
CDX-I0034-M250
102990-192EA
121.85
USD
102990-192
102990-194
102990-196
1-(3-Isothiocyanatobenzyl)-4-[2-(3,4-dihydro-2H-1-benzopyran-6-yl)-5-oxazolyl] pyridinium bromide ≥90% (by HPCE)
1-(3-Isothiocyanatobenzyl)-4-[2-(3,4-dihydro-2H-1-benzopyran-6-yl)-5-oxazolyl]pyridinium bromide
Class of highly photostable, water soluble fluorescent labels with large Stoke′s shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient (Epsilon∼
Formula:
C₂₅H₂₀BrN₃O₂S Storage Temperature: Freezer |
CAS Number:
155863-01-7 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Yellow crystalline. |
Solubility | Soluble in chloroform. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...