9-(Hydroxymethyl)-10-carbamoylacridan ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: 9-HMCA

CDX-H0118-M010 CDX-H0118-M025
102990-128EA 157.85 USD
102990-128 102990-130
9-(Hydroxymethyl)-10-carbamoylacridan ≥98%
9-(Hydroxymethyl)-10-carbamoylacridan

Major intermediate in carbamazepine metabolism. This carbamazepine metabolite can be used as a reference compound.

Formula: C₁₅H₁₄N₂O₂
Storage Temperature: Freezer
CAS Number: 68011-71-2

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (HPLC)
Appearance/Color White powder.
Solubility Soluble in THF and chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR