1-Hydroxy-1,2-benziodoxol-3-one ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: 1-Hydroxy-3-oxo-1,3-dihydro-1,2-benzoiodoxole, IBA

CDX-H0103-M100 CDX-H0103-M500
102990-094EA 210.46 USD
102990-094 102990-096
1-Hydroxy-1,2-benziodoxol-3-one ≥98%
1-Hydroxy-1,2-benziodoxol-3-one

Heterocyclic iodane with catalytic activity in the hydrolytic cleavage of reactive phosphates, phosphonates and carboxylates. Used as catalyst for the micellar cleavage of activated esters and phosphates and shown to be powerful reagent for the cleavage of phosphates in an aqueous cetyltrimethylammonium chloride (CTAC) micellar medium.

Formula: C₇H₅IO₃
MW: 264.1 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01658875
CAS Number: 131-62-4

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White to off-white powder.
Solubility Soluble in toluene.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR