Heptenophos ≥98%

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: Ragadan

CDX-H0095-M100 CDX-H0095-M500
102990-066EA 210.46 USD
102990-066 102990-068
Heptenophos ≥98%
Heptenophos

Synthetic insecticides derived from phosphoric acid, and capable of disrupting neurotransmission in insects and vertebrates. Ectoparasitocidal drug that works as an acetylcholine esterase inhibitor in arthropods. Compound can be used as analytical reference material.

Formula: C₉H₁₂ClO₄P
Storage Temperature: Freezer
MDL Number: MFCD00055303
CAS Number: 23560-59-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Light yellow to very dark yellow solid.
Solubility Soluble in DMF, DMSO or dioxane.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR