N-Decanoyl-L-homoserine lactone ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

CDX-D0332-M005 CDX-D0332-M050
102989-782EA 88.62 USD
102989-782 102989-784
N-Decanoyl-L-homoserine lactone ≥98% (by NMR)
N-Decanoyl-L-homoserine lactone

N-Decanoyl-L-homoserine lactone is a small diffusible signaling molecule involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism. The applications of this molecule include regulation of virulence and exoproteases.

Formula: C₁₄H₂₅NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11113140
CAS Number: 177315-87-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (NMR)
Appearance/Color White powder.
Solubility Soluble in DMSO and dimethylformamide (approx. 30mg/ml).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR