2',7'-Difluorofluorescein ≥95% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
CDX-D0087-M100
CDX-D0087-G001
102987-776EA
253.38
USD
102987-776
102987-778
2',7'-Difluorofluorescein ≥95% (by HPLC)
2',7'-Difluorofluorescein
Fluorescent labelling.
Formula:
C₂₀H₁₀F₂O₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD01311849 CAS Number: 195136-58-4 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or chloroform. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...