N,N-Dimethyl-6-propionyl-2-naphthylamine (Prodan) ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: 6-Propionyl-2-dimethylaminonaphthalene, 1-(6-(Dimethylamino)naphthalen-2-yl)propan-1-one, Prodan, 2-(Dimethylamino)-6-propionylnaphthalene

CDX-D0077-M025 CDX-D0077-M050 CDX-D0077-M500
102987-750EA 153.69 USD
102987-750 102987-752 102987-754
N,N-Dimethyl-6-propionyl-2-naphthylamine (Prodan) ≥98%
N,N-Dimethyl-6-propionyl-2-naphthylamine (Prodan)

N,N-dimethyl-6-propionyl-2-naphthylamine (prodan) has both an electron-donor and an electron-acceptor substituent, resulting in a large excited-state dipole moment and extensive solvent polarity-dependent fluorescence shifts. When prodan is incorporated into membranes, their fluorescence spectra are sensitive to the physical state of the surrounding phospholipids. In membranes, prodan appears to localize at the surface, although Fourier transform infrared measurements indicate some degree of penetration into the lipid interior. Excited-state relaxation of prodan is sensitive to the nature of the linkage between phospholipid hydrocarbon tails and the glycerol backbone. Tubulin and its hydrophobic surfaces have been probed with the enviroment-sensitive probes prodan. Prodan is also used for the generation of peroxy-oxalate chemiluminescence with H2O2.

Formula: C₁₅H₁₇NO
Storage Temperature: Freezer
MDL Number: MFCD00056615
CAS Number: 70504-01-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Light yellow powder.
Solubility Soluble in methanol, DMF, acetonitrile or acetone.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR