CCVJ 97

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 9-(2-Carboxy-2-cyanovinyl)julolidine, (E)-2-Cyano-3-(1,2,3,5,6,7-hexahydropyrido[3,2,1-ij]quinolin-9-yl)acrylic acid

CDX-C0085-M010 CDX-C0085-M125
102989-454EA 235.38 USD
102989-454 102989-456
CCVJ 97
CCVJ

Fluorescent probe for binding to proteins etc. quantum yield increases by decreasing free rotation. Fluorescent molecular rotor.

Formula: C₁₆H₁₆N₂O₂
MW: 268.31 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00274408
CAS Number: 142978-18-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (HPLC)
Appearance/Color Red crystalline solid.
Solubility Soluble in DMSO or chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR