5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester (CDCFDA) 96
Supplier: ADIPOGEN CORP MS
Synonyms:
Carboxy-2′,7′- dichlorofluorescein diacetate, succinimidyl ester, 5 and 6 isomers, Carboxy-DCFDA-N-succinimidyl ester, CDCFDA
CDX-C0059-M050
CDX-C0059-M250
102989-440EA
342
USD
102989-440
102989-442
5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester (CDCFDA) 96
5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester (CDCFDA)
5-(and-6)-Carboxy-2',7'-dichlorofluorescein diacetate, succinimidyl ester is a useful fluorescent tracer that can passively diffuse into cells and covalently label intracellular proteins, resulting in long-term cell labeling. The reagent itself is colorless and nonfluorescent but becomes brightly green fluorescent once it is hydrolyzed by intracellular esterases.
Formula:
C₂₉H₁₇Cl₂NO₁₁ MW: 626.35 g/mol Storage Temperature: Freezer |
CAS Number:
147265-60-9 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in DMSO or DMF. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...