5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE) ≥90% (by HPLC)

Supplier: ADIPOGEN CORP MS

Synonyms: 5-Carboxytetramethylrhodamine N-succinimidyl ester5-TAMRA, SE5-TAMRA-N-succinimidyl esterTAMRA SE, 5 isomer

CDX-C0057-M100 CDX-C0057-M001 CDX-C0057-M010
102987-650EA 3072.46 USD
102987-650 102987-646 102987-648
5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE) ≥90% (by HPLC)
5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE)

Amine-reactive form of 5-carboxytetramethylrhodamine single isomer. Conjugates yield bright, pH-insensitive orange-red fluorescence (Ex/Em: ~546/579nm) with good photostability.

Formula: C₂₉H₂₅N₃O₇
MW: 527.52 g/mol
Storage Temperature: Freezer
CAS Number: 150810-68-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (HPLC)
Appearance/Color Dark violet powder.
Solubility Soluble in DMSO or DMF.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR