5-ROX ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 5-ROX, 5-Carboxy-X-rhodamine

CDX-C0004-M020 CDX-C0004-M200
102987-518EA 213.23 USD
102987-518 102987-520
5-ROX ≥95%
5-ROX

5-ROX is the purified isomer of 5(6)-ROX. It is preferred for some complicated biological applications where reproducibility is really critical. The minor positional difference between 5-ROX and 6-ROX might significantly affect some biological properties of the underlying conjugates. Used for oligonucleotide labeling and automated DNA sequencing applications.

Formula: C₃₃H₃₀N₂O₅
Storage Temperature: Refrigerator
MDL Number: MFCD28355897
CAS Number: 216699-35-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Green solid.
Solubility Soluble in methanol, DMSO or DMF.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR