N-Biotinyl-NH-(PEG)2-COOH DIPEA ≥95% (by HPLC)
Supplier: ADIPOGEN CORP MS
Synonyms:
O-(N-Biotinyl-3-aminopropyl)-O'-(N-glutaryl-3-aminopropyl)-diethyleneglycol . DIPEA
Functionalized biotin. Biotinylated polyethylene glycol (PEG-Biotin) is an avidine or streptavidin reactive PEG derivative that can be used to modify biomolecues or other surfaces through biotin-streptavidin interaction. Used for addition of biotin-PEG to the N-terminus of resin-bound peptides. Can be introduced using standard coupling methods, such as PyBOP or TBTU and has better solubility in DMF and NMP than comparable non-PEG-containing derivatives, such as biotin-Ahx and biocytin.
Formula:
C₂₅H₄₄N₄O₈S·C₈H₁₉N MW: 689.9 g/mol Storage Temperature: Freezer |
CAS Number:
1205744-09-7 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White to off-white powder. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...