Di-tert-butylcyclopentadiene ≥95% (by GC)

Supplier: ADIPOGEN CORP MS
Danger

CDX-B0136-L005 CDX-B0136-L025
102989-392EA 171.69 USD
102989-392 102989-394
Di-tert-butylcyclopentadiene ≥95% (by GC)
Di-tert-butylcyclopentadiene

Building block for synthesis. Frequently used as precursors to di-tert-butylcyclopentadienyl (Cpt) ligand in many sandwich and half sandwich complexes. Di-tert-butylcyclopentadiene reacts with excess potassium tert-butoxide in the presence of elemental selenium to yield cyclopentadienes bridged by two or three selenium atoms through one or two diselenoketal functionalities. It reacts with excess potassium tert-butoxide in the presence of elemental sulfur to yield cyclopentadienes bridged by sulfur atoms via one or two dithioketal functionalities.

Formula: C₁₃H₂₂
Storage Temperature: Freezer
MDL Number: MFCD00674508
CAS Number: 73046-16-9
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Source/Host Synthetic
Purity 95% (GC)
Appearance/Color Transparent to yellow oil.
Solubility Soluble in organic solvents.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR