7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ≥90%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: AMCA, 2H-1-Benzopyran-3-acetic acid, 7-Amino-4-methylcoumarin-3-acetic acid, Aminomethyl coumarin acetic acid

CDX-A0009-M100 CDX-A0009-M500 CDX-A0009-G002
102989-110EA 84.46 USD
102989-110 102989-112 102989-114
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ≥90%
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H)

AMCA is one of the brightest amine-reactive blue fluorescent dyes useful for immunofluorescence and fluorescent labeling (Ex/Em: 353/455nm). It is quite photostable, and its fluorescence is pH-independent from pH 4 to 10. The properties include a relatively large Stoke's shift and resistance to photobleaching. Reactive AMCA derivatives are used as contrasting probes for double and triple labeling in immunofluorescence microscopy, arrays and in situ hybridization.


NHS-AMCA and Sulfo-NHS-AMCA are reactive towards primary amine groups on antibodies, proteins, peptides and other biomolecules. AMCA-Hydrazide is used to label glycosylation sites or for conjugation to carboxyl groups using the crosslinker EDC.

Formula: C₁₂H₁₁NO₄
Storage Temperature: Freezer
MDL Number: MFCD00081051
CAS Number: 106562-32-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (HPLC)
Appearance/Color Brown solid.
Solubility Soluble in DMSO or DMF.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR