Rubrofusarin ≥98%

Supplier: ADIPOGEN CORP MS
Warning

BVT-0395-M001 BVT-0395-M005
102988-954EA 152.31 USD
102988-954 102988-956
Rubrofusarin ≥98%
Rubrofusarin

Monomer of aurofusarin. Mycotoxin. Anticancer and antimycobacterial compound in vivo. Moderate tyrosinase inhibitor. Anti-estrogenic.

Formula: C₁₅H₁₂O₅
MW: 272.3 g/mol
Storage Temperature: Freezer
CAS Number: 3567-00-8

Order Now

Specification Test Results

Source/Host Isolated from Fusarium graminearum.
Purity 98% (HPLC)
Appearance/Color Orange needles.
Solubility Soluble in DMSO. Sparingly soluble in usual organic solvents.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR