19-O-Acetylchaetoglobosin A ≥98%

Supplier: ADIPOGEN CORP MS
Warning

BVT-0383-M001 BVT-0383-M005
102988-934EA 131.54 USD
102988-934 102988-936
19-O-Acetylchaetoglobosin A ≥98%
19-O-Acetylchaetoglobosin A

Cytochalasan alkaloid, derivative of chaetoglobosin A . Mycotoxin. Phytotoxin.

Formula: C₃₄H₃₈N₂O₆
Storage Temperature: Refrigerator
CAS Number: 50939-69-0

Order Now

Specification Test Results

Source/Host Isolated from Chaetomium sp.
Purity 98% (HPLC, NMR)
Appearance/Color Yellow solid.
Solubility Soluble in DMSO, acetone or dichloromethane. Fairly soluble in water or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR