Nebularine ≥98%, high purity
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
9-beta-D-Ribofuranosyl-9H-purine, (2R,3S,4R,5R)-2-(Hydroxymethyl)-5-(9H-purin-9-yl)tetrahydrofuran-3,4-diol
Nucleoside analog. Cytotoxic. DNA, RNA and protein synthesis inhibitor. Shows anticancer activity. Anti-mycobacterial. Has tuberculostatic activity. Antifungal activity. Antiviral (anti-Herpes). Adenosine deaminase inhibitor. Can be used to analyze structure determinants of DNA that are recognized by DNA repair enzymes, to locate triple helices at G-C sequences or as an universal base, which can bind to all four of the nucleosides of DNA.
Formula:
C₁₀H₁₂N₄O₄ MW: 252.2 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00022819 CAS Number: 550-33-4 |
Specification Test Results
Source/Host | Isolated from Streptomyces sp. |
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in DMSO or methanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...