Pimprinine ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 3-(2-Methyl-5-oxazolyl)-1H-indole

BVT-0297-M001 BVT-0297-M005
102988-918EA 180 USD
102988-918 102988-920
Pimprinine ≥98%
Pimprinine

Indole alkaloid. Monoamine oxidase inhibitor (MAOI). Antiepileptic (anticonvulsant) compound. Inhibitor of platelet aggregation and thromboxane A2 synthesis. Weak antifungal and antituberculosis activity. Useful as lead structure for potent antifungal substances.

Formula: C₁₂H₁₀N₂O
MW: 198.2 g/mol
Storage Temperature: Freezer
CAS Number: 13640-26-1

Order Now

Specification Test Results

Source/Host Isolated from Streptomyces sp.
Purity 98% (HPLC, NMR)
Appearance/Color Pale yellow to beige powder.
Solubility Soluble in DMSO, methanol or acetone.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR