Me7BIO ≥95% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 1-Methyl-7-bromo-indirubin-3'-oxime

AG-MR-C0022-M005 AG-MR-C0022-M025
102988-850EA 512.31 USD
102988-850 102988-848
Me7BIO ≥95% (by NMR)
Me7BIO

N-methylated inactive control compound for 7BIO . Aryl hydrocarbon receptor (AhR) agonist; causes redistribution of AhR to the nucleus.

Formula: C₁₇H₁₂BrN₃O₂
MW: 370.2 g/mol
Storage Temperature: Freezer
CAS Number: 916440-98-7

Order Now

Specification Test Results

Purity 95% (NMR)
Appearance/Color Dark red solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR