PLX4720 ≥99% (by HPLC)

Supplier: ADIPOGEN CORP MS

Synonyms: PLX-4720

AG-CR1-3577-M005 AG-CR1-3577-M025
102988-618EA 373.85 USD
102988-618 102988-616
PLX4720 ≥99% (by HPLC)
PLX4720

Potent and selective cell permeable B-Raf (V600E) and c-Raf-1 (Y340D/Y341D) inhibitor. Antitumor compound. ERK phosphorylation is potently inhibited exclusive in B-Raf(V600E)-bearing tumor cell lines.

Formula: C₁₇H₁₄ClF₂N₃O₃S
MW: 413.8 g/mol
Storage Temperature: Freezer
MDL Number: MFCD14635203
CAS Number: 918505-84-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 99% (HPLC)
Appearance/Color Off-white solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR