BQ-788 sodium salt ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS

AG-CP3-0012-C500 AG-CP3-0012-M001
102988-466EA 207.69 USD
102988-466 102988-468
BQ-788 sodium salt ≥95% (by HPLC)
BQ-788 sodium salt

Potent and selective endothelin B receptor (ETBR) antagonist.

Formula: C₃₄H₅₀N₅O₇·Na
Storage Temperature: Freezer
MDL Number: MFCD00797882
CAS Number: 156161-89-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO, 0.1M sodium bicarbonate or 100% ethanol. Slightly soluble in methanol. Insoluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR