Rugulosin ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

AG-CN2-0124-C250 AG-CN2-0124-M001
102987-990EA 62.31 USD
102987-990 102987-992
Rugulosin ≥95% (by HPLC)
Rugulosin

Antibiotic


Source/Host: Isolated from Penicillium sp.
Purity: >95% (HPLC)
Appearance/Color: Yellow solid.
Solubility: Soluble in DMSO, ethanol or methanol.


Antibacteriophage. Antiviral. Mycotoxin. DNA replication, transcription and repair inhibitor. Antibacterial. RNA polymerase and ribonuclease H inhibitor. Insecticidal. Cytotoxic. HIV-1 integrase inhibitor. Shows anti-MRSA (methicillin-resistant Staphylococcus aureus) activity. Potential Hsp90 inhibitor through interacting with N-Hsp90.

Storage Temperature: Freezer
CAS Number: 23537-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR