Imatinib Mesylate

Supplier: ADIPOGEN CORP MS

SYN-1046-M001 SYN-1046-M005 SYN-1046-M010 SYN-1046-M050
102991-606EA 59.54 USD
102991-606 102991-608 102991-610 102991-612
Imatinib Mesylate
Imatinib mesylate

Imatinib is a multi-target inhibitor of v-Abl, c-Kit and PDGFR with IC(50) values of 0.6µM, 0.1µM and 0.1µM, respectively.

Formula: C₂₉H₃₁N₇O
MDL Number: MFCD04307699
CAS Number: 220127-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR