Verapamil Hydrochloride

Supplier: ADIPOGEN CORP MS

CDX-V0007-M100 CDX-V0007-G001 CDX-V0007-G005
102991-166EA 42.92 USD
102991-166 102991-168 102991-170
Verapamil Hydrochloride
Verapamil hydrochloride

Verapamil hydrochloride is an alpha-adrenergic receptor (alpha-AR) antagonist and calcium channel protein inhibitor that blocks the L-type Ca2+ channels in smooth and cardiac muscle cells. Verapamil is an antiarrhythmic agent and vasodilator known to reduce the renal clearance of digoxin and induce apoptosis in primary and metastatic colon adenocarcinoma human cell lines in vitro. It has been observed that verapamil can induce currents by itself, presumably by acting on the potassium and chloride leakage. Verapamil has also been used as an inhibitor of drug efflux pump proteins such as Mdr (P-glycoprotein). Verapamil is a substrate of CYP3A4 and CYP2C6. It is also used in fluorescent cell sorting for DNA content, as it blocks efflux of a variety of DNA-binding fluorophores such as Hoechst 33342. Recent research has shown verapamil to be an effective treatment for diabetes in animal models. Verapamil helps treat diabetes by limiting TXNIP expression.

Formula: C₂₇H₃₈N₂O₄
MDL Number: MFCD00055208
CAS Number: 152-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR