4-Methylumbelliferyl butyrate

Supplier: ADIPOGEN CORP MS

Synonyms: 4-Methyl-2-oxo-2H-chromen-7-yl butyrate

CDX-M0011-G005 CDX-M0011-G020 CDX-M0011-G125
102990-358EA 70.62 USD
102990-358 102990-360 102990-362
4-Methylumbelliferyl butyrate
4-Methylumbelliferyl butyrate

Fluorogenic substrate for butyrate esterase.

Formula: C₁₄H₁₄O₄
MDL Number: MFCD00038087
CAS Number: 17695-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR