5(6)-Carboxyfluorescein diacetate

Supplier: ADIPOGEN CORP MS

CDX-C0046-M025 CDX-C0046-M100 CDX-C0046-G001
102987-628EA 66.46 USD
102987-628 102987-630 102987-632
5(6)-Carboxyfluorescein diacetate
5(6)-Carboxyfluorescein diacetate

CFDA is a popular, sensitive, fluorescent substrate for measuring esterase activity in live cells. It is often used in cell viability assays and "Live-Dead" assay systems, especially automated cell viability assays. The cleavage product produced upon enzymatic or chemical hydrolysis of the acetate group(s) is carboxyfluorescein which is well-retained inside live cells, while the substrate CFDA is membrane permeant.

Formula: C₂₅H₁₆O₉
MDL Number: MFCD00467494
CAS Number: 124387-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR