5(6)-Carboxyfluorescein

Supplier: ADIPOGEN CORP MS

Synonyms: 3′,6′-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthene]-ar-carboxylic acid5(6)-FAM

CDX-C0045-G050 CDX-C0045-G005 CDX-C0045-G001
102987-622EA 62.31 USD
102987-622 102987-624 102987-626
5(6)-Carboxyfluorescein
5(6)-Carboxyfluorescein

Carboxyfluorescein is commonly employed as a polar tracer and used for intracellular pH measurements. It has also been used for studying viability of various types of bacteria.

Formula: C₄₂H₂₄O₁₄
MW: 752.63 g/mol
MDL Number: MFCD00151081
CAS Number: 72088-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR