(+)-Bicuculline

Supplier: ADIPOGEN CORP MS

CDX-B0239-M025 CDX-B0239-M500 CDX-B0239-G005
102989-420EA 99.69 USD
102989-420 102989-422 102989-424
(+)-Bicuculline
(+)-Bicuculline

GABAA receptor antagonist.

Formula: C₂₀H₁₇NO₆
MDL Number: MFCD00005006
CAS Number: 485-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR