2,5-Bis(4-aminophenyl)-1,3,4-oxadiazole

Supplier: ADIPOGEN CORP MS

CDX-B0219-M020 CDX-B0219-M250
102989-410EA 139.85 USD
102989-410 102989-412
2,5-Bis(4-aminophenyl)-1,3,4-oxadiazole
2,5-Bis(4-aminophenyl)-1,3,4-oxadiazole

Fluorescent stain for DNA.

Formula: C₁₄H₁₂N₄O
MDL Number: MFCD00042667
CAS Number: 2425-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR